missing translation for 'onlineSavingsMsg'
Learn More

Ibidi Culture-Insert 3 Well in μ-Dish 35 mm, High ibiTreat

Product Code. 16471661
Encompass_Preferred
Click to view available options
No. of Wells:
2
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 16471661

Brand: Ibidi 80366

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Culture-Insert 3 Well in μ-Dish 35 mm, High ibiTreat is a 3 well silicone insert with 2 defined cell-free gaps of 500 μm.

  • A 3 well silicone insert with 2 defined cell-free gaps of 500 μm
  • suitable for wound healing
  • migration assays
  • 2D invasion assays
  • and co-cultivation of cells
  • in a μ-Dish 35 mm high with #1.5 Polymer Coverslip bottom
  • tissue culture treated
  • sterilized

Specifications

Type Culture Insert
Quantity 30
Wells 2
Sterility Sterile
No. of Wells 2
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.