Learn More
Thermo Scientific™ Culti-Loops™ Sensititre™ MIC Breakpoint Plates / Just One Strips QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4657020
Additional Details : Weight : 0.01000kg ??product.attributeSet.Transport?? : UN number : 3373 Chem class : 6.2
Description
Thermo Scientific™ Culti-Loops™ Sensititre MIC Breakpoint Plates/Just One Strips Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Enterococcus faecalis ATCC™ 29212™†
Escherichia coli ATCC™ 25922™†
Escherichia coli ATCC™ 35218™†
Pseudomonas aeruginosa subsp. ATCC™ 27853™†
Staphylococcus aureus subsp. aureus ATCC™ 29213™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
29212TM* | |
Culti-Loops | |
Organism Sets | |
5 Loops/Package |
Sensititre™ MIC Breakpoint Plates; Just One Strips QC Set/5 | |
Culti-Loops QC Set | |
Qualitative QC | |
5 Loops/Organism |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.