Learn More
Thermo Scientific™ Culti-Loops™ Sensititre™ MIC Breakpoint Plates / Just One Strips QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4657020
Additional Details : Weight : 0.01000kg ??product.attributeSet.Transport?? : UN number : 3373 Chem class : 6.2
Product Code. 15249287
  • 635.00 € / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Culti-Loops™ Sensititre MIC Breakpoint Plates/Just One Strips Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Enterococcus faecalis ATCC™ 29212™†
Escherichia coli ATCC™ 25922™†
Escherichia coli ATCC™ 35218™†
Pseudomonas aeruginosa subsp. ATCC™ 27853™†
Staphylococcus aureus subsp. aureus ATCC™ 29213™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
29212TM*
Culti-Loops
Organism Sets
5 Loops/Package
Sensititre™ MIC Breakpoint Plates; Just One Strips QC Set/5
Culti-Loops QC Set
Qualitative QC
5 Loops/Organism
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.