Learn More
Thermo Scientific™ Culti-Loops™ Campylobacter jejuni ATCC® 33560™
Description
Confidently carry out testing of media, reagents, and identification & antimicrobial susceptibility kits and evaluate bacteriological procedures with ready-to-use Thermo Scientific™ Culti-Loops™ QC organisms.
Culti-Loops™ Campylobacter jejuni ATCC® 33560™ is recommended to be used for antimicrobial susceptibility quality control testing.
Campylobacter jejuni ATCC® 33560™ is recommended to be used for antimicrobial susceptibility quality control testing by CLSI and EUCAST.
Technique:
The loop may be dissolved in a liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000. Alternatively, it can be streaked directly onto appropriate media.
Includes:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | ATCC™ 33560™ |
| Description | Campylobacter jejuni ATCC™ 33560™ |
| Format | Culti-Loops |
| Target Organism Class | Gram-negative, Helical-shaped bacteria |
| Test Type | Qualitative QC |
| Quantity | 5 Loops/Pk |
| Strain Designations | NCTC 11351, CIP 70.2T, DSM 4688, CCUG 11284 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.