Learn More
Thermo Scientific™ Culti-Loops™ API™ RapID™ 20E QC Set
Description
Thermo Scientific™ Culti-Loops™ API RapID 20E Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedure.
Includes:
Escherichia coli ATCC™ 11775™†
Klebsiella pneumoniae subsp.pneumoniae ATCC™ 35657™†
Proteus hauseri ATCC™ 13315™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifications
Specifications
| ATCC® | 11775TM* |
| Description | API Rapid 20E QC Set/3 |
| Format | Culti-Loops |
| Product Type | Culti-Loops QC Set |
| Target Organism Class | Organism Sets |
| Test Type | Qualitative QC |
| Packaging Type | 5 Loops/Pack |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.