missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Beryllium Copper, CDA 1751, C17510, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Beryllium Copper, CDA 1751, C17510, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Zinc/Silver/Tin/Lead/Silicon/Cobalt/Aluminium/Iron/Beryllium/Nickel/Copper |
| CAS | 7440-50-8 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.0009 %/Zinc 0.001 %/Silver 0.0011 %/Tin 0.0013 %/Lead 0.0015 %/Silicon 0.004 %/Cobalt 0.0084 %/Aluminium 0.0104 %/Iron 0.0108 %/Beryllium 0.404 %/Nickel 1.905 %/Copper 97.61 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction