missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTS-1027, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
CTS-1027 is a potent small molecule inhibitor of MMPs, with IC50s of 0.3 nM, 0.5 nM for MMP2, MMP13, respectively, and has > 1,000 fold selectivity over MMP1.
Specifications
Specifications
| Chemical Name or Material | CTS-1027 |
| CAS | 193022-04-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H20ClNO6S |
| Quantity | 50 mg |
| Synonym | Ro 1130830 RS 130830 |
| Solubility Information | DMSO : ≥ 100 mg/mL (234.81 mM) |
| SMILES | O=C(NO)C1(CCOCC1)CS(=O)(C2=CC=C(C=C2)OC3=CC=C(C=C3)Cl)=O |
| Molecular Weight (g/mol) | 425.88 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction