missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CtIP Antibody (19E8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-41205
This item is not returnable.
View return policy
Description
CtIP Monoclonal specifically detects CtIP in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay.
Specifications
| CtIP | |
| Monoclonal | |
| 1.3mg/mL | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Proximity Ligation Assay, Chromatin Immunoprecipitation (ChIP) | |
| RBBP8 | |
| Human CtIP | |
| Immunogen affinity purified | |
| RUO | |
| 5932 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay | |
| 1900000000 | |
| Unconjugated | |
| CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog | |
| Mouse | |
| 125 kDa | |
| 100 μL | |
| Primary | |
| This antibody will recognize human CtIP. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction