missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CST 626
GREENER_CHOICE

Product Code. 30000663
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000663 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000663 Supplier Tocris Bioscience™ Supplier No. 8021/2
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent pan IAP Degrader (PROTAC™)

CST 626 is a potent IAP Degrader (PROTAC™) (DC50 values at 16 hours are 0.7 nM, 2.4 nM and 6.2 nM at XIAP, cIAP1 and cIAP2 respectively; Dmax = 90-99%). Reduces cell viability in some cell types, including SUDHL6 and MOLM13 (IC50 values are 16 nM and 21 nM respectively).

PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S,4S)-1-((S)-2-Cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)-4-(3-((5-(((S)-1-((2S,4R)-4-hydroxy-2-(((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-5-oxopentyl)oxy)phenoxy)-N-((R)-1,2,3,
Quantity 2 mg
Target Active Degraders
Molecular Formula C61H82N8O9S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.