missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CSNK1A1L Recombinant Protein

Brand:  Abnova™ H00122011-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137513

  • 508.00 € / 25µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH28723
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
62.81
8
Glutathione Sepharose 4 Fast Flow
25 ug
MTNNSGSKAELVVGGKYKLVRKIGSGSFGDVYLGITTTNGEEVAVKLESQKVKHPQLLYESKLYTILQGGVGIPHMHWYGQEKDNNVLVMDLLGPSLEDLFNFCSRRFTMKTVLMLADQMISRIEYVHTKNFLHRDIKPDNFLMGTGRHCNKLFLIDFGLAKKYRDNRTRQHIPYREDKHLIGTVRYASINAHLGIEQSRRDDMESLGYVFMYFNRTSLPWQGLKAMTKKQKYEKISEKKMSTPVEVLCKGFPAEFAMYLNYCRGLRFEEVPDYMYLRQLFRILFRTLNHQYDYTFDWTMLKQKAAQQAASSSGQGQQAQTQTGKQTEKNKNNVKDN
MGC33182/RP11-532O21.2
CSNK1A1L
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
122011
CSNK1A1L (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CSNK1A1L
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions