missing translation for 'onlineSavingsMsg'
Learn More

CSN1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18639701 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18639701 0.02 mL 0.02mL
18627102 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18639701 Supplier Novus Biologicals Supplier No. NBP2929070.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CSN1 Polyclonal antibody specifically detects CSN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSN1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias COP9 signalosome complex subunit 1, COPS1JAB1-containing signalosome subunit 1, CSN1MGC71287, G protein pathway suppressor 1SGN1, GPS-1, Protein MFH, Signalosome subunit 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 258-527 of human GPS1 (NP_997657.1). VSKAESTPEIAEQRGERDSQTQAILTKLKCAAGLAELAARKYKQAAKCLLLASFDHCDFPELLSPSNVAIYGGLCALATFDRQELQRNVISSSSFKLFLELEPQVRDIIFKFYESKYASCLKMLDEMKDNLLLDMYLAPHVRTLYTQIRNRALIQYFSPYVSADMHRMAAAFNTTVAALEDELTQLILEGLISARVDSHSKILYARDVDQRSTTFEKSLLMGKEFQRRAKAMMLRAAVLRNQIHVKSPPREGSQGELTPANSQSRMSTNM
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuronal Cell Markers, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2873
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.