missing translation for 'onlineSavingsMsg'
Learn More

CSH1 Monoclonal Antibody (OTI3C8), TrueMAB™, OriGene

Product Code. 17516322
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17516322 30 μL 30µL
17596312 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17516322 Supplier Origene Technologies Supplier No. TA506793S

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CSH1 Monoclonal antibody specifically detects CSH1 in Human samples. It is validated for Western Blot

Specifications

Antigen CSH1
Applications Western Blot
Classification Monoclonal
Clone OTI3C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CSH1
Gene Accession No. P0DML2
Gene Alias Choriomammotropin; chorionic somatomammotropin A; chorionic somatomammotropin hormone 1; chorionic somatomammotropin hormone 1 (placental lactogen); Chorionic somatomammotropin hormone 2; chorionic somatomammotropin-1; CS-1; CSA; CSH1; CSH2; CSMT; GHB3; growth hormone B3; hCS-1; hCS-A; hPL; Lactogen; PL; placental lactogen
Host Species Mouse
Immunogen Full length human recombinant protein of human CSH1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1442
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.