Learn More
Crystal Ponceau 6R (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | Acid Red 44 |
| CAS | 2766-77-0 |
| Molecular Formula | C20 H12 N2 O7 S2 . 2 Na |
| Quantity | 5 g |
| Synonym | 1,3-Naphthalenedisulfonic acid, 7-hydroxy-8-(1-naphthalenylazo)-, disodium salt (9CI), 2-Naphthol-6,8-disulfonic acid, 1-(1-naphthylazo)-, disodium salt (6CI), C.I. Acid Red 44 (7CI), C.I. Acid Red 44, disodium salt (8CI), Acid Leather Ponceau 6R, Acid Ponceau 6R, Acid Red 6A, Acidal Crystals Ponceau, Brilliant Crystal Scarlet, C.I. 16250, Colacid Red 6A, Crystal Ponceau, Crystal Ponceau 6R, Crystal Ponceau M 6R, Crystal Scarlet, Crystal Scarlet 6R, Crystal Tertracid Ponceau 6R, Kemacid Leather Ponceau 6R, Ponceau 6R, Ponceau 6R Crystals, Ponceau Cristallise Extra A Export, Ponceau Crystals 6R, Romexal Red 6A, Triacid Ponceau 6R |
| InChI Formula | InChI=1S/C20H14N2O7S2.2Na/c23-17-9-8-13-10-14(30(24,25)26)11-18(31(27,28)29)19(13)20(17)22-21-16-7-3-5-12-4-1-2-6-15(12)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].Oc1ccc2cc(cc(c2c1N=Nc3cccc4ccccc34)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC Name | disodium;7-hydroxy-8-[(E)-naphthalen-1-yldiazenyl]naphthalene-1,3-disulfonate |
| Molecular Weight (g/mol) | 502.43 |
| Formula Weight | 501.9881 |