Description
- Formulated to allow supercooling of the sample in the cooling bath and maintain a constant bath temperature of −7.0°C ± 0.5°C
- Used as the temperature control liquid in all cryoscope baths.
- Manufactured for use with all cryoscopes compliant to International Reference Standard ISO5764/IDF108 for the determination of freezing point in milk.
Specifications
Specifications
| Description | Liquid format in HDPE Bottle GL45 |
| For Use With (Equipment) | Cryoscope bath |
| Quantity | 500 mL |