Learn More
Cryoscope Bath Liquid, Reagecon
BIOPHARMA
Product Code. 11523122 Shop All Reagecon Products
Change view
Click to view available options
For Use With (Equipment):
Cryoscope bath
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11523122 Cryoscope bath 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11523122 Supplier Reagecon Supplier No. CRYBL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Temperature control liquid for cryoscope baths.
  • Formulated to allow supercooling of the sample in the cooling bath and maintain a constant bath temperature of −7.0°C ± 0.5°C
  • Used as the temperature control liquid in all cryoscope baths.
  • Manufactured for use with all cryoscopes compliant to International Reference Standard ISO5764/IDF108 for the determination of freezing point in milk.
Specifications
Description Liquid format in HDPE Bottle GL45
For Use With (Equipment) Cryoscope bath
Quantity 500 mL