Learn More
Globe Scientific Cryogenic Vial Cap Inserts
Product Code. 31923933
Change view
Click to view available options
Color:
Assorted
Blue
Gray
Green
Lavender
Orange
Pink
Red
White
Yellow
Unit Size:
Pack of 1000
Pack of 600
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
31923933 Orange Pack of 1000
31922557 Gray Pack of 1000
31922661 Assorted Pack of 600
31923446 Blue Pack of 1000
31923359 Green Pack of 1000
31922826 Pink Pack of 1000
31922860 Red Pack of 1000
31921726 Lavender Pack of 1000
31922567 White Pack of 1000
31923480 Yellow Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 31923933 Supplier Globe Scientific Supplier No. 30079N
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
Ideal for color coding and sample identification.
  • Made from polypropylene
  • Nest easily into place on both internal and external caps
Specifications
Color Orange
Material Polypropylene
Sterility Non-sterile