Learn More
TPP™ Cryo Tubes with Cryogenic Rack
Product Code. 11331794
Change view
Click to view available options
:
Cryogenic Rack with Cryo Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11331794 Cryogenic Rack with Cryo Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11331794 Supplier TPP™ Supplier No. 99016
Please to purchase this item. Need a web account? Register with us today!
Single-handed operation is possible using TPP™ Cryogenic Rack with Cryo Tubes
  • Star-shaped insertion system in grooves allows one hand operation when used with star foot TPP cryo tubes
  • Features non-slip properties
  • Ready-mounted rubber feet
Specifications
Dimensions (L x W x H) 100 x 200 x 25 mm
Quantity 12/cs.
Material Polycarbonate
Type Cryo Tubes with Cryogenic Rack