Learn More
TPP™ Cryo Tubes
Description
TPP™ Cryo Tubes are used to store samples in freezers or similar low-temperature equipment.
- External thread
- Colour-coding inserts in a range of colours
- Abstain from using silicone gasket, enabling secure and tight closure of tube
- Puncture and tear resistant re-closable bags with zipper and laser perforation
- Self-standing, star foot
- Graduated with large writing surface
Specifications
Specifications
| Product Type | Cryogenic Storage Tube |
| Description | Dimensions: 12 x 48 mm |
| Capacity (Metric) | 1.5 mL |
| Material | Polypropylene |
| No. per Case | 800 |