missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Cryo.s Round Bottom 4 mL Polypropylene Tube with External Thread Cap
Ideal for storage of cell cultures, tissue samples, microbiological samples, material of human or animal origin as well as antibodies, RNA, DNA and protein samples.
Brand: Greiner Bio-One 127261
Additional Details : Weight : 20.00000kg
Description
- With or without ID writing area
- With Star foot base
- Sterile
- Non-pyrogenic and non-toxic
- Includes 100 white cap inserts
- Polyethylener case
- Dia. x Height: 12.4 x 83mm
Service Plan
No
Specifications
Cryogenic Storage Tube | |
Natural | |
Round | |
External | |
83 mm | |
Sterile |
12.4 mm | |
4 mL | |
Polypropylene | |
Starfoot Base and 150 white cap inserts | |
Polyethylene case | |
Threaded |