Learn More
Greiner Bio-One Cryo.s Round Bottom 2 mL Polypropylene Tubes with Internal Thread Cap
GREENER_CHOICE
Product Code. p-7075343
Click to view available options
Closure Color:
Natural
Bottom Shape:
Round
Round (Starfoot)
Style:
ID Writing Area
No Writig Area
No Writing Area
Thread Style:
External
Internal
Unit Size:
Pack of 500
Product Code. 10727611
Brand: Greiner Bio-One 121261
Please to purchase this item. Need a web account? Register with us today!
Made of high-grade polypropylene which guarantees high thermal resistance.
  • Internal thread has silicone gasket
  • With ID writing area
  • Sterile
  • Non-pyrogenic and non-toxic
  • Includes 150 white cap inserts
  • Polyethylener case
  • With or without star foot base
  • Dia. x Height: 12
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens. Contents non-cytotoxic
Closure Color Natural
Autoclavable Not Autoclavable
Capacity (Metric) 2 mL
Bottom Shape Round
Material Polypropylene
Thread Style External
Includes 150 white cap inserts
No. per Case 500
No. per Pack 100
Style No Writing Area
Temperature Range (Metric) -196°C to +121°C
Volume (Metric) Working 1.8 to 2.0 mL
Sterility Sterile
Closure Material Polypropylene
Closure Type Screw Cap