Learn More
Greiner Bio-One Cryo.s Freezing Tubes with Datamatrix Coding
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic, endotoxin-free
- High thermal resistance (196°C to +121°C)
- Laser-written ECC200 Datamatrix coding
- Strong resistance to mechanical and ch
Compliance
CE
Specifications
Specifications
| Product Type | Vial |
| Closure Color | Natural |
| Bottom Shape | Conical |
| Material | Polyethylene |
| Thread Style | Internal |
| Includes | Cap inserts for labeling |
| Volume (Metric) Working | 1 mL |
| Sterility | Sterile |
| Closure Type | Screw Cap |