Learn More
Tempshield™ Cryo-Industrial Gauntlet Fabric Gloves
Product Code. 11813861
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
1 pair
Product Code. 11813861
Brand: Tempshield™ CIGXWP
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Tempshield™ Cryo-Industrial™ Gauntlet Fabric Gloves protect your hands and arms when working in hazardous, ultra-cold environments.
  • Durable industrial safety gloves for protection of hands and arms when working in hazardous, ultra-cold environments on heavy-duty applications
  • Designed to provide a high level of thermal protection, abrasion resistance, flexibility and dexterity
  • Manufactured from multi-layered, state-of-the-art materials
  • Designed to wick away moisture for comfort when being used for extended periods of time
  • Not intended for actual immersion in liquid nitrogen or other cryogenic liquids
Specifications
Color Blue
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gauntlet
Size X-Large
Sterility Non-sterile
Length (Metric) 430 to 475 mm