Learn More
Tempshield™ Cryo-Industrial Gauntlet Fabric Gloves
Click to view available options
Size:
Medium
Large
X-Large
Unit Size:
1 pair
Description
- Durable industrial safety gloves for protection of hands and arms when working in hazardous, ultra-cold environments on heavy-duty applications
- Designed to provide a high level of thermal protection, abrasion resistance, flexibility and dexterity
- Manufactured from multi-layered, state-of-the-art materials
- Designed to wick away moisture for comfort when being used for extended periods of time
- Not intended for actual immersion in liquid nitrogen or other cryogenic liquids
Specifications
Specifications
| Color | Blue |
| Material | Fabric |
| Powdered | Powder Free |
| Product Type | Cryogenic Gauntlet |
| Size | X-Large |
| Sterility | Non-sterile |
| Length (Metric) | 430 to 475 mm |