Learn More
CRX Mouse anti-Human, Unconjugated, Clone: 1C3C11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67555-1-IG-150UL
 View more versions of this product
Product Code. 16890246
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The CRX antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRX. This antibody recognizes human, mouse, rat, pig antigen. The CRX antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CRX
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
1C3C11
Unconjugated
150 μL
1406