Learn More
CRX Mouse anti-Human, Unconjugated, Clone: 1C3C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67555-1-IG-150UL
Description
The CRX antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRX. This antibody recognizes human, mouse, rat, pig antigen. The CRX antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CRX | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 1C3C11 | |
| Unconjugated | |
| 150 μL | |
| 1406 | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.