missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Plays important functions in early cell signaling. Binds the cAMP response element (CRE) (consensus: 5′-GTGACGT[AG][AG]-3′), a sequence present in many viral and cellular promoters. Activator of the NF-ELAM1/delta-A site of the E-selectin promoter. Has no intrinsic transcriptional activity, but activates transcription on formation of JUN or FOS heterodimers. Also can bind TRE promoter sequences when heterodimerized with members of the JUN family. Isoform 4/ATF-A0acts as a domit repressor of the E-selectin/NF-ELAM1/delta-A promoter. Isoform 5/ATF-4acts as a negative regulator, inhibiting both ATF2 and ATF7 transcriptional activities. It may exert these effects by sequestrating in the cytoplasm the Thr-53 phosphorylating kinase, preventing activation.
Specifications
Specifications
| Antigen | CRKRS |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | CDK12 |
| Gene Accession No. | Q9NYV4 |
| Gene Alias | CDC2 related protein kinase 7, CDK12, CRK7, CRKR, CRKRS, KIAA0904 |
| Gene Symbols | CDK12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?