Learn More
CRISPR-Cas9 Antibody (6H4), PE, R&D Systems™
Product Code. 18614760 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
18614760 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18614760 Supplier Novus Biologicals Supplier No. NBP281124PE
  • ONLINE EXCLUSIVE PRICE
    631.17 €
    553.35 € / 0.1mL
    Save 77.82 €
    12% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Mouse Monoclonal Antibody
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Applications Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot
Product Type Antibody
Product Line Novus