Learn More
Microspec™ Cribbar Sample Bottles - Sterile Na2S2O3 Dosed
Cribbar Sample Bottles are sterile Na2S2O3 dosed
Brand: Microspec™ P9
Additional Details : Weight : 3.90000kg
Description
- Cribbar Sample Bottles
- Sterile Na2S2O3 Dosed
- 18 mg/L
Specifications
Natural | |
UK | |
250 mL |
18 mg/L | |
Sterile |