missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRHR1/CRF1 Antibody (343919), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3930F
This item is not returnable.
View return policy
Description
CRHR1/CRF1 Monoclonal antibody specifically detects CRHR1/CRF1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| CRHR1/CRF1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 343919 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor | |
| NS0 mouse myeloma cell line transfected with human CRHR-1, Ser24-Val415, Accession # NP_004373 | |
| 0.1 mL | |
| GPCR, Neuroscience, Neurotransmission | |
| 1394 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction