missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CREB3 Polyclonal antibody specifically detects CREB3 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CREB3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000 |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | basic leucine zipper protein, cAMP responsive element binding protein 3, cAMP responsive element binding protein 3 (luman), cAMP-responsive element-binding protein 3, CREB-3, cyclic AMP response element (CRE)-binding protein/activating transcriptionfactor 1, cyclic AMP-responsive element-binding protein 3, LUMAN, LZIPMGC15333, MGC19782, Transcription factor LZIP-alpha |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human CREB3 (NP_006359.3). MELELDAGDQDLLAFLLEESGDLGTAPDEAVRAPLDWALPLSEVPSDWEVDDLLCSLLSPPASLNILSSSNPCLVHHDHTYSLPRETVSMDLESESCRKEGTQMTPQHMEELAEQEIARLVLTDEEKSLLEKEGLILPETLPLTKTEEQILKRVRRKIRNKRSAQESRRKKKVYVGGLESRVLKYTAQNMELQNKVQLLEEQNLSLLDQLRKLQAMVIEISNKTSSSSTC |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?