missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase, Muscle/CKMM Antibody (CKMM/3341), PE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Creatine Kinase, Muscle/CKMM Monoclonal antibody specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | Creatine Kinase, Muscle/CKMM |
| Applications | ELISA, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | CKMM/3341 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK |
| Host Species | Mouse |
| Immunogen | Recombinant full-length Creatine Kinase, Muscle/CKMM protein (Uniprot: P06732) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?