missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Kinase B Mouse anti-Human, Clone: 8.00E+04, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Creatine Kinase BB Monoclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Creatine Kinase B |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 80000 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1063, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Formulation | Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide |
| Gene Alias | B-CK, Creatine kinase B chain, creatine kinase, brain, creatine kinase-B, EC 2.7.3, EC 2.7.3.2 |
| Gene Symbols | CKB |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?