Learn More
Abeba™ Crawler ALU 4570 Shoes
Product Code. 15651608
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy
Product Code. 15651608
Brand: Abeba™ 4570/40
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Aluminum toe cap provides lightweight toe protection. Sanitized™ treated.
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather with breathable textile inlays
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
  • Scratch-resistant coating in toe and heel area
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRC
Color White
Material Leather
Size 40
Sole Features Slip-resistant
Sole Material Polyurethane
Style 4570
Toe Material Aluminum