Learn More
Edmund Bühler GmbH™ Cramp Strip
Click to view available options
For Use With (Equipment):
Combifix
KL and KS shakers
SM Shakers
Unit Size:
Each
Description
- Keeps horizontal separating funnels in place
- Includes clamps for NS 19 and NS 29
Specifications
Specifications
| Product Type | Cramp Strip |
| For Use With (Application) | For fastening horizontal separating funnels |
| For Use With (Equipment) | SM Shakers |
| Includes | 5 Clamps for NS 19, 2 clamps for NS 29 |
| No. of Positions | 7 |