missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRALBP Antibody (OTI1G6), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71275APC
This item is not returnable.
View return policy
Description
CRALBP Monoclonal specifically detects CRALBP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| CRALBP | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| RLBP1 | |
| Full length recombinant protein of human CRALBP (NP_000317) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI1G6 | |
| Flow Cytometry | |
| Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1 | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 6017.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction