missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPSF7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16517381
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16517381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16517381 Supplier Invitrogen™ Supplier No. PA589500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LIDIYADEEF NQDPEFNNTD QIDLYDDVLT ATSQPSDDRS SSTEPPPPVR QEPSPKPNNK TPAILYTYSG LRNRRAAVYV GSFSWWTTDQ QLIQVIRSIG VYDVVELKFA ENRANGQSKG YAEVVVASEN SVHKLLELLP GKVLNGEKVD VRPATRQNLS QFEAQARKRE CVRVPRGGIP PRAHSRDSSD S; Positive Samples: Jurkat, HepG2, Mouse thymus; Cellular Location: Nucleus.

FLJ12529 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPSF7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CPSF7
Gene Accession No. Q8BTV2, Q8N684
Gene Alias 5730453I16Rik; AL022757; C330017N18Rik; CFIm59; cleavage and polyadenylation specific factor 7; cleavage and polyadenylation specific factor 7, 59kDa; cleavage and polyadenylation specificity factor 59 kDa subunit; Cleavage and polyadenylation specificity factor subunit 7; cleavage factor Im complex 59 kDa subunit; CPSF 59 kDa subunit; Cpsf7; FLJ12529; LOW QUALITY PROTEIN: cleavage and polyadenylation specificity factor subunit 7; pre mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor I, 59 kDa subunit; pre-mRNA cleavage factor Im 59 kDa subunit; RGD1305441; Unknown (protein for IMAGE:7949905)
Gene Symbols CPSF7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-240 of human FLJ12529 (NP_0790873).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269061, 79869
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.