missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15489-1-AP-150UL
This item is not returnable.
View return policy
Description
CPSF6 is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitement of other processing factors. The cleavage factor complex is composed of four polypeptides.The protein encoded by this gene is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitement of other processing factors. The cleavage factor complex is composed of four polypeptides. This gene encodes the 68kD subunit. It has a domain organization reminiscent of spliceosomal proteins.Specifications
| CPSF6 | |
| Polyclonal | |
| Unconjugated | |
| CPSF6 | |
| CFIM, CFIM68, CPSF 68 kDa subunit, CPSF6, HPBRII 4, HPBRII 7, Protein HPBRII 4/7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11052, 299811, 432508 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16630, Q6NVF9 | |
| Cpsf6 | |
| CPSF6 Fusion Protein Ag7852 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction