missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPS1 Antibody (CPS1/8417), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23839-20ug
This item is not returnable.
View return policy
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CPS1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 | |
| Recombinant human CPS1 protein | |
| 20 μg | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 1373 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CPS1/8417 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction