missing translation for 'onlineSavingsMsg'
Learn More

CPI-455, MedChemExpress

Product Code. 16412586
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412586

Brand: Medchem Express HY100421

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CPI-455 is a specific, pan-KDM5 inhibitor with an IC50 of 10 nM for KDM5A. CPI-455 mediates KDM5 inhibition, elevates global levels of H3K4me3, and decreases the number of drug-tolerant persister cancer cells in multiple cancer cell line models treated with standard chemotherapy or targeted agents.

CPI-455 is a specific, pan-KDM5 inhibitor with an IC50 of 10 nM for KDM5A. CPI-455 mediates KDM5 inhibition, elevates global levels of H3K4me3, and decreases the number of drug-tolerant persister cancer cells in multiple cancer cell line models treated with standard chemotherapy or targeted agents.

Specifications

Chemical Name or Material CPI-455
Physical Form Solid
CAS 1628208-23-0
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C16H14N4O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 34 mg/mL (122.17 mM)
SMILES N#CC1=C2NC(C3=CC=CC=C3)=C(C(C)C)C(N2N=C1)=O
Molecular Weight (g/mol) 278.31
Formula Weight 278.31
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.