Learn More
CPC™ Sanitary AseptiQuik Insert
Product Code. 30968744
Change view
Click to view available options
Maximum Pressure:
45 psig
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Maximum Pressure Unit Size
30968744 45 psig Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30968744 Supplier CPC™ Supplier No. AQC44012
  • ONLINE EXCLUSIVE PRICE
    281.00 €
    269.25 € / Pack of 5
    Save 11.75 €
    4% Off
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Sterility Sterile
Maximum Pressure 45 psig
Max. Operating Temperature (Metric) 40°C
Product Type Sanitary Insert