Learn More
Cole-Parmer™ CPC™ Panel-mount Gasket for Miniature Non-spill Connectors
Click to view available options
Packaging Quantity:
10/Pk.
Unit Size:
Pack of 10
Description
- Less than 0.025mL of spillage.
- Unique valve design for ultra-low air inclusion in line.
- Non-spill protection and high-flow fluid processing with a small footprint.
- Economical alternative for non-spill solutions.
- This coupling features an innovative twist-to-connect design with an integrated locking mechanism for secure coupling engagement and worry-free fit.
- Both halves of the coupling include shut-off valves to guarantee minimal spillage when disconnected.
- The same shut-off valves ensure less than 0.009mL air inclusion to minimize contaminants in the process line.
- This slim, compact coupling reduces valuable space required for the connection and minimizes components in your process.
Specifications
Specifications
| Type | Connector |
| Packaging Quantity | 10/Pk. |
| Description | Quick-Disconnect |
| Material | Buna N |