Learn More
Tocris Bioscience™ CP 465022 hydrochloride
GREENER_CHOICE
Product Code. 18094692
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18094692
Brand: Tocris Bioscience™ 2932/10
Please to purchase this item. Need a web account? Register with us today!
Selective, non-competitive AMPA antagonist
Selective, non-competitive AMPA antagonist (IC50 = 25 nM in rat cortical neurons) that displays potent anticonvulsant activity. Also significantly blocks the persistent component of Nav1.6 channel activity. Brain penetrant and orally active.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1785666-59-2
Molecular Formula C26H25Cl2FN4O
Specifications
Quantity 10 mg
Formula Weight 499.41
Purity >99%
Molecular Weight (g/mol) 499.411
Chemical Name or Material CP 465022 hydrochloride