missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX5b Antibody, CoraFluor™ 1, Novus Biologicals™
Description
COX5b Polyclonal antibody specifically detects COX5b in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | COX5b |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | COXVB, Cytochrome c oxidase polypeptide Vb, cytochrome c oxidase polypeptide VB, mitochondrial, cytochrome c oxidase subunit 5B, mitochondrial, cytochrome c oxidase subunit Vb |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human COX5b (Accession#: P10606; Ala32-His129) |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?