Learn More
Ibidi Coverslips for Sticky-Slides Uncoated
Click to view available options
:
Coverslips
Description
- Polymer Coverslips #1.5
- Uncoated
- 25 mm x 75 mm
- hydrophobic
- non-sterile
Specifications
Specifications
| Product Type | Coverslip |
| Length (Metric) | 75 mm |
| Width (Metric) | 25 mm |
| Sterility | Sterile |
| Quantity | 100 |