missing translation for 'onlineSavingsMsg'
Learn More

Ibidi Coverslips und Coverslip Pick-Up Tool for 3 Well Chamber, Removable

Coverslips und Coverslip Pick-Up Tool for 3 Well Chamber, Removable are coverslips #1.5H (170 μm +/- 5 μm) D 263 M Schott glass.

Brand:  Ibidi 10815

Additional Details : Weight : 1.00000kg

Product Code. 15643404

  • 152.46 € / Pack of 100
Estimated Shipment: 11-07-2024
to see stock.
Add to basket

Description

Description

  • Coverslips #1.5H (170 μm +/- 5 μm) D 263 M Schott glass
  • Ø 15 mm
  • non-sterile
  • including tool for handling
Specifications

Specifications

Coverslip
25 mm
100
75 mm
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers