missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, MedChemExpress

Product Code. p-200101222
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16489126

Brand: Medchem Express HYN0709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Coumarin is the primary bioactive ingredient in Radix Glehniae, named Beishashen in China, which possesses many pharmacological activities, including anticancer, anti-inflammation and antivirus activities.

Coumarin is the primary bioactive ingredient in Radix Glehniae, named Beishashen in China, which possesses many pharmacological activities, including anticancer, anti-inflammation and antivirus activities.

Specifications

Chemical Name or Material Coumarin
Physical Form Solid
CAS 91-64-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H6O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (684.28 mM) ∣H2O : 4 mg/mL (27.37 mM; Need ultrasonic)
SMILES O=C1C=CC2=CC=CC=C2O1
Molecular Weight (g/mol) 146.14
Formula Weight 146.14
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.