Learn More
Coumarin-7-pinacolboronate, TRC
Product Code. 30352228
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30352228 10 mg
30363142 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30352228 Supplier Toronto Research Chemicals Supplier No. TRCC75540010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 190788-61-5
Molecular Formula C15 H17 B O4
Molecular Weight (g/mol) 272.1
IUPAC Name 7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)chromen-4-one
SMILES CC1(C)OB(OC1(C)C)c2ccc3C(=O)C=COc3c2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C15H17BO4/c1-14(2)15(3,4)20-16(19-14)10-5-6-11-12(17)7-8-18-13(11)9-10/h5-9H,1-4H3
Formula Weight 272.122
Chemical Name or Material Coumarin-7-pinacolboronate