Learn More
Invitrogen™ Cortisol Monoclonal Antibody (F4P1A3)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514676
Description
MA5-14676 targets Cortisol in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MA5-14676 immunogen is cortisol-3-CMO-BSA. Product MA514676 is a smaller package size of MIC0201 (formerly sold as a Seradyn product).
Cortisol is the most potent glucocorticoid produced by the human adrenal. It is synthesized from cholesterol and its production is stimulated by pituitary adrenocorticotropic hormone (ACTH) which is regulated by corticotropin releasing factor (CRF). ACTH and CRF secretions are inhibited by high cortisol levels in a negative feedback loop. In plasma a majority of cortisol is bound with high affinity to corticosteroid binding globulin (CBG or transcotin). Cortisol acts through specific intracellular receptors and affects numerous physiologic systems including immune function, glucose counter regulation, vascular tone, and bone metabolism.
Specifications
| Cortisol | |
| Monoclonal | |
| 1 mg/mL | |
| 0.01M potassium phosphate with 1 mg/mL BSA, 50% glycerol, 2.5 mM KCl, 0.135M NaCl and 0.05% sodium azide | |
| Cortisol-3-CMO-BSA. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid | |
| Chemical |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| F4P1A3 | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.