Learn More
Masterflex™ Corrugated FEP Tubing
Click to view available options
Diameter (Metric) Inner:
5.6 mm
Unit Size:
Each
Description
- Corrugations prevent kinking and help tubing retain shape without limiting pressure or vacuum abilities.
- Custom extruded.
- Can be cut to desired length.
- Sold in 5-ft lengths.
- Alternating sections of corrugated (10-in. length), and noncorrugated (2-in. length), with noncorrugated 1-in. lengths at both ends.
- Accepts compression fittings.
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;NSF-61;USP;3A;European Pharmacopoeia;NSF-51 |
| Diameter (Metric) Inner | 5.6 mm |
| Material | FEP |
| Diameter (Metric) Outer | 6.4 mm |
| Overall Length (Metric) | 1.5 m |
| Thickness (Metric) | 0.41275 mm |
| Min. Operating Temperature (Metric) | -184°C |
| Color | Translucent |
| Maximum Pressure | 250 psi |
| Vacuum Level | Withstands 27-in. Hg vacuum |
| Show More |