Description
- Tear-resistant, foil laminated pouches with 3 x 4 in. white marking label
- Heat-sealable
- Nonsterile
Specifications
Specifications
| Description | For 25 Slide Storage Box |
| Quantity | 50/Cs. |
| Color | Gray |
| Type | Storage Pouch |
| For Use With (Application) | Printed microarrays |