Learn More
Corning™ LSE™ Vortex Mixer Accessory, Processing Heads
Heads allow for processing of larger numbers of tubes
Brand:  Corning™ 480103
Additional Details : Weight : 0.21772kg
 View more versions of this product
Product Code. 11931981
  • 47.11 € / Each
Estimated Shipment: 04-10-2024
to see stock.
Add to basket
Description
Description
This is an optional head for processing larger numbers of tubes, for use with Corning™ LSETM Vortex Mixer.
Specifications
Specifications
8 x 15 mL tubes, 8 x 12 mm dia. tubes, and 8 x 13 mm dia. tubes
LSE Vortex Mixer