Learn More
(-)-Corey lactone diol, MedChemExpress
Product Code. 16449336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16449336 Supplier Medchem Express Supplier No. HYW008393
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress (-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis.
(-)-Corey lactone diol is a reduced version of corey aldehyde. A building block in the chemical synthesis.
Specifications
Chemical Name or Material (-)-Corey lactone diol
CAS 32233-40-2
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C8H12O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (580.79 mM; Need ultrasonic)
SMILES OC[C@@H]([C@H](O)C[C@]1([H])O2)[C@@]1([H])CC2=O
Molecular Weight (g/mol) 172.18
Formula Weight 172.18
Percent Purity 97.0%
Grade Research
Physical Form Solid