missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Technical, Metal Foil, (about 0.1mm), Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
Specifications
Specifications
| Color | Brown |
| Melting Point | 1083°C |
| Boiling Point | 2595°C |
| Quantity | 250 g |
| Packaging | Plastic container |
| Form | Solid |
| Formula Weight | 63.54g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction