missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Brass Majors, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Brass Majors, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Bismuth/Arsenic/Tin/Aluminium/Lead/Iron/Nickel/Manganese/Zinc/Copper/Silicon |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0056 %/Bismuth 0.0116 %/Arsenic 0.012 %/Tin 0.0127 %/Aluminium 0.0148 %/Lead 0.0188 %/Iron 0.182 %/Nickel 0.212 %/Manganese 2.84 %/Zinc 36.67 %/Copper 60.03 %/Silicon (0.032) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction